Anti-CHRM2 Rabbit Monoclonal Antibody

GTX62624
89364-534EA 556.26 CAD
89364-534
Anti-CHRM2 Rabbit Monoclonal Antibody
The muscarinic cholinergic receptors belong to a larger family of G protein-coupled receptors. The functional diversity of these receptors is defined by the binding of acetylcholine to these receptors and includes cellular responses such as adenylate cyclase inhibition, phosphoinositide degeneration, and potassium channel mediation. Muscarinic receptors influence many effects of acetylcholine in the central and peripheral nervous system. CHRM2 (Muscarinic acetylcholine receptor M2) is involved in the mediation of bradycardia and causes a decrease in cardiac contractility.

Type: Primary
Antigen: CHRM2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR