Anti-CIB1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX100849
89348-712EA 881.1 CAD
89348-712
Anti-CIB1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to CIB1 (calcium and integrin binding 1 (calmyrin))

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CIB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR