Anti-CIDE-3 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX70476
89365-004EA 907.62 CAD
89365-004
Anti-CIDE-3 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal to human CIDE - 3

Western blot - Use at 1:500-1,000 dilution. However, the investigator should determine the optimal dilution for a specific application such as immunohistochemistry.

Type: Primary
Antigen: CIDE-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR