Anti-CKR2 Rabbit Polyclonal Antibody

GTX77805
89366-308EA 516.95 CAD
89366-308
Anti-CKR2 Rabbit Polyclonal Antibody
Receptors for the C-C chemokine family include CKR-I, CKR-2A, CKR-3, CKR-4, CKR- 5 and the Duffy blood group antigen. The C-C receptors are important in the function of T cell chemotaxis and migration of phagocytic cells to sites of inflammation.

Type: Primary
Antigen: CKR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR