Anti-CLK1 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX107766
89355-136EA 881.1 CAD
89355-136
Anti-CLK1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to CLK1 (CDC-like kinase 1)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CLK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR