Anti-Cofilin 2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX14133
89359-954EA 990.03 CAD
89359-954
Anti-Cofilin 2 Rabbit Polyclonal Antibody
Antibodies
Cofilin is a widely distributed intracellular actin-modulating protein that binds and depolymerizes filamentous F-actin and inhibits the polymerization of monomeric G-actin in a pH-dependent manner. Cofilin 2 is the muscle isoform.

WB: Use at a concentration of 1 ?g/ml. Detects a band of approximately 21 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Cofilin 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR