Anti-Compound S Rabbit Polyclonal Antibody

Supplier: Genetex
GTX10995
89356-122EA 553.14 CAD
89356-122
Anti-Compound S Rabbit Polyclonal Antibody
Compound S Levels Normal plasma cortisol <1000 pg/0.1 mlAfter administration of2-3 grams of metyrapone 7,000 - 25,000 pg/0.1ml

Type: Primary
Antigen: Compound S (11 DEOXYCORTISOL)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR