Anti-CORIN Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX46410
89363-606EA 937.18 CAD
89363-606
Anti-CORIN Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to CORIN (C-terminal)

Recommended Dilutions: For Western Blot: Use at 2.5 ug/ml.<p>Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: CORIN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR