Anti-Cortisol Rabbit Polyclonal Antibody

GTX10998
89356-132EA 543.72 CAD
89356-132
Anti-Cortisol Rabbit Polyclonal Antibody
Cortisol LevelsNormal plasma cortisol(8 am) 12,800 "" 4,000 pg/0.1mlHyperadrenalism(e.g., severe operation) 60,000 pg/0.1mlHypoadrenalism(e.g., Addisonis disease,pituitary insufficiency) <1000 pg/0.1ml

Type: Primary
Antigen: Cortisol
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR