Anti-Cortisol Mouse Monoclonal Antibody [clone: XM210]

Supplier: GeneTex
GTX21949
89351-696EA 643 CAD
89351-696
Anti-Cortisol Mouse Monoclonal Antibody [clone: XM210]
Antibodies
Cortisol is the most potent glucocorticoid produced by the human adrenal.

Type: Primary
Antigen: Cortisol
Clonality: Monoclonal
Clone: [XM210]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR