Anti-COX6A2 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX108261
89355-420EA 691.89 CAD
89355-420
Anti-COX6A2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to COX6A2 (cytochrome c oxidase subunit VIa polypeptide 2)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: COX6A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR