Anti-CSF1R Rat Monoclonal Antibody

GTX72220
89352-976EA 516.68 CAD
89352-976
Anti-CSF1R Rat Monoclonal Antibody
CD115, also known as c-fms, macrophage Colony Stimulating Factor Receptor (MCSFR), or CSF-1R, exists in immature (gp130 c-fms) and mature (gp150 c-fms) forms. CSF-1R exhibits an intrinsic tyrosine-specific protein kinase activity.

Type: Primary
Antigen: CSF1R
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR