Anti-CSK Goat Polyclonal Antibody

Supplier: GeneTex
GTX89784
89367-016EA 874.26 CAD
89367-016
Anti-CSK Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to CSK

Approx 50kDa band observed in Jurkat lysate (predicted MW of 50.7kDa according to NP_004374.1). Recommended for use at 0.5-2µg/ml. In paraffin embedded Human Colon shows staining of a cell layer between the mucosal columns and the muscularis mucosae. Recommended concentration, 3-6µg/ml.

Type: Primary
Antigen: CSK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR