Anti-CSRP3 Chicken Polyclonal Antibody

GTX14013
89367-230EA 677.25 CAD
89367-230
Anti-CSRP3 Chicken Polyclonal Antibody
Cysteine and glycine-rich protein 3 (CSRP3) is a member of the CSRP family of genes encoding a group of LIM domain proteins, which may be involved in regulatory processes important for development and cellular differentiation. The LIM/double zinc-finger motif found in CRP3 is found in a group of proteins with critical functions in gene regulation, cell growth, and somatic differentiation. Very strongly similar to rat MLP; contains zinc-finger LIM domains. Other genes in the family include CSRP1 and CSRP2.

For WB: Use at an assay dependent dilution. Predicted molecular weight: 22 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: CSRP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR