Anti-CTBP1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX14411
89359-996EA 994.72 CAD
89359-996
Anti-CTBP1 Rabbit Polyclonal Antibody
Antibodies
The phosphoprotein C-terminal binding protein 1 (CtBP1) binds the C-terminus of adenovirus E1A protein. CtBP1 is a transcriptional repressor and may play a role during cellular proliferation.

ICC: Recommended dilution: 1?g/ml. IP: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CTBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR