Anti-CUL1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX22964
89351-836EA 565.01 CAD
89351-836
Anti-CUL1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Cullin 1

  • Immunoprecipitation (native): use at a concentration of 10μg / mg (protein A). Western Blotting: use at a concentration of 5 - 10μg / ml for 2hrs at RT. Immunohistochemistry (formalin fixed paraffin embedded): use at a concentration of 5 - 10μg / ml for 30 min at RT. Not tested in other applications. Optimal dilutions / concentrations should be determined by the end user. By Western blot, this antibody detects a band of 86 kDa, which corresponds to the predicted molecular weight of Cullin 1. No special pretreatment is required for immuno histochemical staining of formalin fixed tissues.

Type: Primary
Antigen: CUL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR