Anti-CUL2 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX21870
89351-658EA 581.69 CAD
89351-658
Anti-CUL2 Rabbit Polyclonal Antibody
Cullin 2 is a member of the family of human Cullin genes (CUL1, 2, 3, 4a, 4b and 5) homologous to the S. cerevisiae cdc53 gene. CUL 1 forms a complex with human p19skp1 and F box protein p45skp2, together with cdc53, as three subunits, form a ubiquitin ligase controlling proteolysis of G1 cell cycle regulatory proteins. In contrast to CUL 1, CUL 2 does not associate with p19skp1 and p45skp2. CUL 2 forms a complex with an inactive transcriptional elongation complex, SIII, formed by three subunits: Elongain C, Elongin B and VHL.

Type: Primary
Antigen: CUL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR