Anti-CXCL11 Chicken Polyclonal Antibody

GTX82411
89368-082EA 666.36 CAD
89368-082
Anti-CXCL11 Chicken Polyclonal Antibody
Chemotactic for interleukin-activated T-cells but not unstimulated T-cells. neutrophils or monocytes. Induces calcium release in activated T-cells. Binds to CXCR3. May play an important role in CNS diseases which involve T-cell recruitment. May play a role in skin immune responses. Chemokines are a group of small (approximately 8 to 14 kD). mostly basic. structurally related molecules that regulate cell trafficking of various types of leukocytes through interactions with a subset of 7-transmembrane. G protein-coupled receptors. Chemokines also play fundamental roles in the development. homeostasis. and function of the immune system. and they have effects on cells of the central nervous system as well as on endothelial cells involved in angiogenesis or angiostasis. Chemokines are divided into 2 major subfamilies. CXC and CC. This protein is a CXC member of the chemokine superfamily. Its encoded protein induces a chemotactic response in activated T-cells and is the dominant ligand for CXC receptor-3. The gene encoding this protein contains 4 exons and at least three polyadenylation signals which might reflect cell-specific regulation of expression. IFN-gamma is a potent inducer of transcription of this gene.

Type: Primary
Antigen: CXCL11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR