Anti-CXCR4 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX70529
89365-034EA 727.83 CAD
89365-034
Anti-CXCR4 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal to Human CXC Chemokine Receptor 4 ( CXCR4 )

For IHC-P: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: CXCR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR