Anti-CXCR5 Mouse Monoclonal Antibody [clone: 51505.111]

GTX10405
89350-210EA 548.15 CAD
89350-210
Anti-CXCR5 Mouse Monoclonal Antibody [clone: 51505.111]
Chemokines have been sub-divided into families on the basis of the relative position of their cysteine residues. The alpha and beta families, with four cysteine residues, are the largest and best characterized. In the alpha family, one amino acid separates the first two cysteine residues (CXC); in the beta family the two cysteine residues (CC) are adjacent to each other. The alpha chemokines that contain the N-terminal Glu-Leu-Arg amino acid sequence (ELR-motif) are chemotactic for neutrophils (such as IL8), while those that do not, act on lymphocytes (such as IP10 and MIG). Examples of chemokines under the beta family category are MCP1-5 and RANTES. The chemokine lymphotactin belongs to the gamma family, with only two cysteines (C), and the recently described fractalkine or neurotactin is a member of the delta family and has the first two cysteine residues separated by three amino-acids (CXXXC). Chemokines bind to specific G protein-coupled cell surface receptors on target cells. Five CXC receptors (CXCR1-5), nine CC receptors (CCR1-9) and one CXXXC receptor (CX3CR1) have been cloned to date. Expression of chemokine receptors can be restricted to some cell types (CXCR1 is expressed in neutrophils) while others (such as CCR2) are expressed in a wide variety of cells. Receptor expression has also been found to be constitutive (including down regulation), inducible or restricted to a cell state of activation. In addition, some chemokine receptors are also expressed in non-hematopoietic cells, such as nerve, endothelial and epithelial cells. This suggests that chemokines have other roles besides leucocyte chemotaxis. CX3CR1, for example, is highly expressed in adult brain. Chemokine receptors are linked to phospholipases through the Gi class of G proteins (inhibition by pertussis toxin). Receptor activation leads to a cascade of cellular events including generation of inositol triphosphate, calcium release and activation of protein kinase C. Chemokine receptors also activate small GTP-binding proteins of the Ras and Rho families, the latter being involved in cell motility events. In addition, chemokines bind to non-signaling molecules such as the Duffy antigen receptor for chemokines (DARC) which may act to remove chemokines from the circulation, and heparan sulfates proteoglycans which may serve to establish an ECM concentration gradient. Naive B cells express CXCR5, which has been shown by gene targeting experiments to be essential for the formation of certain follicles and germinal centers. The human CXCR5 ligand, B-cell attracting chemokine (BCA1, BLC), and its mouse homologue, B lymphocyte chemoattractant (BLC), 4 are produced by cells in follicles and may play a role in organizing their structure by attracting naive B cells as well as activated Th (T helper) cells.

Type: Primary
Antigen: CXCR5
Clonality: Monoclonal
Clone: 51505.111
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR