Anti-CXCR6 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX77937
89366-432EA 728.03 CAD
89366-432
Anti-CXCR6 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to CXC Chemokine Receptor 6 (CXCR6)

Type: Primary
Antigen: CXCR6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR