Anti-Cyanine Sheep Polyclonal Antibody

GTX26902
89362-254EA 556.8 CAD
89362-254
Anti-Cyanine Sheep Polyclonal Antibody
Type: Primary
Antigen: Cyanine
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR