Anti-CCNF Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX103027
89349-824EA 653.46 CAD
89349-824
Anti-CCNF Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to CCNF (cyclin F)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CCNF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR