Anti-COX2 Rabbit Monoclonal Antibody

GTX62145
89364-326EA 585.93 CAD
89364-326
Anti-COX2 Rabbit Monoclonal Antibody
Cytochrome c oxidase subunit 2 (CO2 or MTCO2) is 1 of 3 mitochondrial DNA (mtDNA) encoded subunits (MTCO1, MTCO2, MTCO3) of respiratory Complex IV. Complex IV is located within the mitochondrial inner membrane and is the third and final enzyme of the electron transport chain of mitochondrial oxidative phosphorylation. It collects electrons from ferrocytochrome c (reduced cytochrome c) and transfers them to oxygen to give water. The energy released is used to transport protons across the mitochondrial inner membrane. Complex IV is composed of 13 polypeptides. Subunits 1, 2 and 3 are encoded by the mtDNA while the remaining subunits are nuclear encoded .

Type: Primary
Antigen: COX2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR