Anti-CYP2D6 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX113667
89357-680EA 881.1 CAD
89357-680
Anti-CYP2D6 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Cytochrome P450 2D6 (cytochrome P450, family 2, subfamily D, polypeptide 6)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CYP2D6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR