Anti-Cytokeratin (Keratin Hair Cortex) Mouse Monoclonal Antibody

Supplier: GeneTex
GTX16113
89360-172EA 1029.5 CAD
89360-172
Anti-Cytokeratin (Keratin Hair Cortex) Mouse Monoclonal Antibody
Antibodies
The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. Most of the type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains and are clustered in a region on chromosome 17q21.2.

Type: Primary
Antigen: Cytokeratin (Keratin Hair Cortex)
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR