Anti-FAK Rabbit Monoclonal Antibody

GTX61768
89364-172EA 556.26 CAD
89364-172
Anti-FAK Rabbit Monoclonal Antibody
Focal adhesion kinase (FAK) is a non-receptor protein-tyrosine kinase implicated in signaling pathways involved in cell motility, proliferation and apoptosis . FAK is composed of a central catalytic domain flanked by large N- and C-terminal regions. FAK is activated by phosphorylation at tyrosine 397 in response to integrin clustering which can be induced by cell adhesion or antibody cross-linking or via G-protein-coupled receptor (GPCR) occupancy by ligands such as bombesin or lysophosphatidic acid . Phosphorylation of FAK Tyr-397 creates a binding site for Src-family kinases, and phosphorylation of FAK Tyr-576/Tyr-577 in the kinase domain activation loop enhances catalytic activity . Increased FAK expression has been correlated with the enhanced motility and invasiveness of human tumor cells, as well as with promoting increased cell proliferation .

Type: Primary
Antigen: FAK (Phospho TYR576/577)
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR