Anti-IgG2 Mouse Monoclonal Antibody [clone: 3C7]

GTX21935
89351-676EA 472.59 CAD
89351-676
Anti-IgG2 Mouse Monoclonal Antibody [clone: 3C7]
ELISA: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Secondary
Antigen: IgG2
Clonality: Monoclonal
Clone: 3C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR