Anti-IgG2a Mouse Monoclonal Antibody

Supplier: GeneTex
GTX80657
89368-020EA 705.06 CAD
89368-020
Anti-IgG2a Mouse Monoclonal Antibody
Antibodies
Mouse IgG2a isotype control

Type: Primary
Antigen: IgG2a
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR