Anti-IgG2 β Rat Monoclonal Antibody

GTX74671
89365-524EA 472.59 CAD
89365-524
Anti-IgG2 β Rat Monoclonal Antibody
Rat IgG2b isotype control

Type: Primary
Antigen: IgG2 β
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR