Anti-IgG Chicken Polyclonal Antibody

GTX82028
89368-038EA 666.36 CAD
89368-038
Anti-IgG Chicken Polyclonal Antibody
IgG is the most abundant class of antibodies found in blood serum and lymph. It is active against bacteria. fungi. viruses. and foreign particles. Immunoglobulin G antibodies trigger action of the complement system.

Type: Primary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR