Anti-IgM Mouse Monoclonal Antibody [clone: 2A6]

Supplier: GeneTex
GTX21946
89351-690EA 638.22 CAD
89351-690
Anti-IgM Mouse Monoclonal Antibody [clone: 2A6]
Antibodies
Human IgM is an antibody that remains in the bloodstream where it can kill bacteria that enter the blood stream. Elevated levels of IgM are associated with Waldenstrom's Macroglobulinemia.

Conjugation: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Can be used for passive haemagglutination. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Secondary
Antigen: IgM
Clonality: Monoclonal
Clone: 2A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR