Anti-Insulin Mouse Monoclonal Antibody

GTX73586
89353-220EA 527.4 CAD
89353-220
Anti-Insulin Mouse Monoclonal Antibody
This antibody recognizes purified insulin from the pancreas of human, bovine, horse, sheep as well as proinsulin from human. Cross reaction has been observed with insulin containing cells in fixed sections of pancreas from human, porcine, dog, rabbit, bovine, sheep, rat, guinea pig and cat.

Type: Primary
Antigen: Insulin
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Cat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR