Anti-Integrin alpha 3 Mouse Monoclonal Antibody

Supplier: GeneTex
GTX11767
89358-794EA 821.69 CAD
89358-794
Anti-Integrin alpha 3 Mouse Monoclonal Antibody
Antibodies
The URO panel of antibodies was developed to immuno-anatomically dissect the nephron from glomerulus to bladder urothelium - these antibodies detect various cell surface antigens specific to segments of the renal tubule. Several have been shown to occur as tissue related markers of renal cancers.This antibody reacts predominantly with adherent cell types and is absent from cells growing in suspension, e.g. leukemia, lymphoma, lymphoblastoid, or other hematopoietic cell lines. In tissues, the URO1 antigen is expressed by glomeruli, the basal layer of transitional epithelium and skin, and the basement membrane of glands. All bladder tumors express URO1, as do most kidney tumors and other epithelial tumors (e.g. colon, ovary, lung and breast). Astrocytoma, melanoma, sarcoma, and lymphoma specimens are negative for URO1 expression.

Type: Primary
Antigen: Integrin alpha 3
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR