Anti-SCGB2A2 Rabbit Polyclonal Antibody

GTX80654
89353-866EA 816.67 CAD
89353-866
Anti-SCGB2A2 Rabbit Polyclonal Antibody
Type: Primary
Antigen: SCGB2A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR