Anti-RNF8 Goat Polyclonal Antibody

Supplier: GeneTex

GTX15850
89360-132EA 748.85 CAD
89360-132
Anti-RNF8 Goat Polyclonal Antibody
Antibodies
The protein encoded by this gene contains a RING finger motif and a FHA domain. This protein has been shown to interact with several class II ubiquitin-conjugating enzymes (E2), including UBE2E1/UBCH6, UBE2E2, and UBE2E3, and may act as an ubiquitin ligase (E3) in the ubiquitination of certain nuclear proteins. Alternatively spliced transcript variants encoding distinct isoforms have been reported.

Approx 55kDa band observed in Human Placenta and Lung lysates (predicted MW of 56kDa according to NP_003949). Recommended for use at 0.1-1µg/ml. This antibody is expected to recognize both reported isoforms (NP_003949.1 and NP_898901.1).

Type: Primary
Antigen: RNF8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR