Anti-ROC1 Rabbit Polyclonal Antibody

GTX77671
89366-232EA 517.99 CAD
89366-232
Anti-ROC1 Rabbit Polyclonal Antibody
ROC1 encodes an evolutionarily conserved protein that interacts with cullins. The protein plays a unique role in the ubiquitination reaction by heterodimerizing with cullin-1 to catalyze ubiquitin polymerization. It also may be involved in the regulation of protein turn-over.

Type: Primary
Antigen: ROC1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR