Anti-MST1R Rabbit Polyclonal Antibody

GTX106254
89354-562EA 258.54 CAD
89354-562
Anti-MST1R Rabbit Polyclonal Antibody
Receptor for macrophage stimulating protein (MSP). Has a tyrosine-protein kinase activity.

Type: Primary
Antigen: MST1R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR