Anti-MAPT Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX86147
89366-900EA 962.74 CAD
89366-900
Anti-MAPT Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Tau (phospho Ser422)

We recommend the following starting dilutions: WB: 1:500~1:1000 IHC: 1:50~1:100 ELISA: 1:10000Optimal dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MAPT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR