Anti-CK19 Rabbit Polyclonal Antibody

GTX73529
89353-200EA 501.26 CAD
89353-200
Anti-CK19 Rabbit Polyclonal Antibody
This antibody reacts with a 40 kD protein. Cytokeratin 19 belongs to type A (acidic) subfamily of low molecular weight cytokeratins. It is expressed in various different human tissues except in liver. Keratin 19 is not expressed in hepatocytes, therefore cytokeratin 19 is useful in the identification of liver metastasis.

Type: Primary
Antigen: CK19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR