Anti-CK19 Mouse Monoclonal Antibody [clone: BA16]

Supplier: GeneTex

GTX40210
89363-134EA 640.88 CAD
89363-134
Anti-CK19 Mouse Monoclonal Antibody [clone: BA16]
Antibodies
Mouse monoclonal [BA16] to Cytokeratin 19

For IHC-Fr:(acetone fixed) Use at an assay dependent dilution. For WB: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: CK19
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR