Anti-CK19 Mouse Monoclonal Antibody [clone: CK 202]

Supplier: GeneTex
GTX82955
89353-878EA 998.26 CAD
89353-878
Anti-CK19 Mouse Monoclonal Antibody [clone: CK 202]
Antibodies
Mouse monoclonal antibody [CK 202] to Cytokeratin 19

IHC, ICC (frozen or formalin–fixed paraffin-embedded (FFPE) tissue sections, cell smears) For IHC dilute conc. antibodies 1:50-1:100, use streptavidin~biotin system or polymer system, incubate 30 minutes at room temperature.

Type: Primary
Antigen: CK19
Clonality: Monoclonal
Clone: CK 202
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR