Anti-CK7 Rabbit Monoclonal Antibody

GTX61887
89364-230EA 556.26 CAD
89364-230
Anti-CK7 Rabbit Monoclonal Antibody
Keratins are cytoplasmic intermediate filament proteins expressed by epithelial cells. Keratin 7 (CK7) is expressed in a wide range of epithelial structures in humans. The expression pattern in mice is similar to the reported human distribution, with substantial expression observed in lung, bladder, mesothelium, hair follicle, and ductal structures . Protein expression levels of CK7 appear significantly higher in chromophobe RCC compared to oncocytoma (p < 0.001). Studies provide further evidence that CK7 and parvalbumin immunostains may be useful in differentiating oncocytoma from chromophobe RCC in problematic cases . Also, the pattern of immunohistochemical expression of CK 7 along with CK 20 is commonly used to assess possible primary sites of metastatic carcinomas .

Type: Primary
Antigen: CK7
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR