Anti-CK8 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX112975
89357-260EA 881.1 CAD
89357-260
Anti-CK8 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Cytokeratin 8 (keratin 8)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CK8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR