Anti-Cytokeratin PAN Mouse Monoclonal Antibody [clone: AE1/AE3]

Supplier: GeneTex
GTX75520
89353-360EA 375.71 CAD
89353-360
Anti-Cytokeratin PAN Mouse Monoclonal Antibody [clone: AE1/AE3]
Antibodies
Is a pan-cytokeratin reagent, being a cocktail of clones AE1 and AE3 which provides the broadest spectrum of reactivity to the 19 known human epidermal keratins. It produces positive staining in virtually all epithelia.

Type: Primary
Antigen: Cytokeratin PAN
Clonality: Monoclonal
Clone: [AE1/AE3]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR