Anti-D(1A) DopamineR (DRD1) Rabbit Monoclonal Antibody

GTX61777
89364-178EA 556.26 CAD
89364-178
Anti-D(1A) DopamineR (DRD1) Rabbit Monoclonal Antibody
D(1A) dopamine receptor (DRD1) belongs to the G-protein coupled receptor 1 family and it is one of the five types of dopamine receptors (D1-D5) . DRD1 is the most expressed dopamine receptor in central nervous system and it is involved in the regulation of neuronal growth & development, and modulation of dopamine receptor D2-mediated events. G protein coupled DRD1 activates adenylyl cyclase which leads to activation of cyclic AMP dependent protein kinases. Polymorphism of DRD1 gene has been implicated in bipolar I disorder and attention deficit hyperactivity disorder (ADHD) .

Type: Primary
Antigen: D(1A) DopamineR (DRD1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR