Anti-DDDDK TAG Goat Polyclonal Antibody

Supplier: GeneTex

GTX21238
89367-054EA 802.74 CAD
89367-054
Anti-DDDDK TAG Goat Polyclonal Antibody
Antibodies
This is a useful tool for the localization and characterization of DDDDK-tagged proteins.

Type: Primary
Antigen: DDDDK TAG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR