Anti-DDR1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX25508
89361-660EA 912.1 CAD
89361-660
Anti-DDR1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to DDR1

Recommended Starting Dilutions: For WB: Use at a dilution of 1:1000. For IHC-P: Use at a dilution of 1:10-1:50. For FACS: Use at a dilution of 1:10-1:50. Optimal concentration should be determined by the end user.

Type: Primary
Antigen: DDR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR