Anti-Ctnnd1 Mouse Monoclonal Antibody [clone: 6H11]

Supplier: GeneTex
GTX11508
89358-242EA 1076.07 CAD
89358-242
Anti-Ctnnd1 Mouse Monoclonal Antibody [clone: 6H11]
Antibodies
delta 1 Catenin is an efficient tyrosine kinase substrate implicated both in cell transformation by SRC and in ligand-induced receptor signaling through the EGF, PDGF, CSF-1 and ERBB2 receptors. The association of catenins to cadherins produces a complex which is linked to the actin filament network, and which seems to be of primary importance for cadherins cell-adhesion properties.

ELISA: Use at an assay dependent dilution. ICC: Use at an assay dependent dilution. IF: Use at a dilution of 1/100. IP: Use at an assay dependent dilution. WB: Use at a dilution of 1/10000. Detects a band of approximately 120 kDa (and a possible doublet band of approximately 200 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Ctnnd1
Clonality: Monoclonal
Clone: 6H11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Chicken, Dog, Rat, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR