Anti-Catenin D2 Rabbit Polyclonal Antibody

GTX11352
89357-590EA 484.4 CAD
89357-590
Anti-Catenin D2 Rabbit Polyclonal Antibody
Delta-Catenin, is a member of the p120 catenin subfamily of catenins. Delta-Catenin/NPRAP was identified as a protein homologous to plakophilin 1 and as a protein interacting with the loop region of presenilin 1 (PS1), the gene most commonly mutated in Alzheimers disease. Delta-Catenin/NPRAP is almost exclusively expressed in the central nervous system mainly during early brain development. Delta-Catenin/NPRAP has been shown to co-localize and interact with N-cadherin in the mouse brain and to undergo dynamic relocalization during brain development.

Type: Primary
Antigen: Catenin D2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR